PTFE Inline Filter
PTFE Inline Filter
Actual product may vary
PTFE Inline Filter
Economical in-line particulate filter maintains the cleanliness and proper operation of your vacuum pump. Welch™ PTFE Inline Filter protects your vacuum pump from fine particulates and aerosols to 0.2 microns.
Have Questions?
Catalog NumberMaterial (Membrane)
FIL112555-04PTFE
Catalog number FIL112555-04
Price (SGD)
-
Material (Membrane):
PTFE
  • Protects vacuum pump from particulates
  • 0.2 micron pore size
  • PTFE filter
  • Polypropylene housing
Specifications
AutoclavableAutoclavable
Material (Membrane)PTFE
Pore Size0.2 μm
For Use With (Application)Particulate filtration, vacuum pump
Housing MaterialPP
Quantity1 Each
TypeInline Filter, DN 8
Unit SizePK