Access Key Options for Ultra-Low Temperature Freezers
Access Key Options for Ultra-Low Temperature Freezers
Actual product may vary
Access Key Options for Ultra-Low Temperature Freezers
Record who opened the ultra low freezer door, when it was opened and how long it was open with access key options.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
FFAC34567
also known as AVL
Thermo Scientific TSU SeriesField installed
FAC34567LETSX Series, Revco RLE, HERAfreeze HLE, Forma 89000, TLE SeriesFactory installed. No stand-alone orders.
Catalog number FFAC34567
also known as AVL
Price (SGD)
2,378.00
EA
Add to cart
For Use With (Equipment):
Thermo Scientific TSU Series
Type:
Field installed
Price (SGD)
2,378.00
EA
Add to cart
Analyze records of which staff opened the freezer door, what time is was opened and for what duration with the Thermo Scientific™ Access Key Options for Ultra-Low Temperature Freezers.
Features
  • Card access comes with 5 key cards
  • Field installed - purchased separately
  • Factory installed in the plant with purchase of unit
Specifications
TypeField installed
For Use With (Equipment)Thermo Scientific TSU Series
IncludesAccess Key Option; Card access control w/five key cards
Unit SizeEA