AREX 5 Digital Hotplate Stirrer
AREX 5 Digital Hotplate Stirrer
Actual product may vary
AREX 5 Digital Hotplate Stirrer
AREX 5 Digital Hotplate Stirrer
AREX 5 Digital Hotplate Stirrer
AREX 5 Digital Hotplate Stirrer
AREX 5 Digital Hotplate Stirrer with CerAlTop plate ensures precise thermoregulation of the media, high safety and maximum reliability.
Have Questions?
Catalog NumberPlug Type
FBV17812027EU/UK
Catalog number FBV17812027
Price (SGD)
782.00
Special offer
Ends: 31-Mar-2026
810.00
Save 28.00 (3%)
Each
Add to cart
Plug Type:
EU/UK
Price (SGD)
782.00
Special offer
Ends: 31-Mar-2026
810.00
Save 28.00 (3%)
Each
Add to cart
PRECISE THERMOREGULATION WITH EXTERNAL TEMPERATURE PROBES AND DIGITAL VTF
  • The ideal solutions for temperature-sensitive applications in many industrial and academic laboratories
  • Precise thermoregulation of the medium with the additional Pt100/Pt1000 probe (± 1.0°C) and VTF digital thermoregulator (± 0.5°C)
  • Thermoregulation range from room temperature up to 300°C.
A SAFE Hotplate STIRRER DESIGNED TO LAST
  • The insulating disk protects the internal electronic parts to resist even in the most challenging environments and applications (IP 42 protection)
  • An illuminated indicator always informs the operator when the heating is on and a Hot Surface Safety message is displayed when the temperature is above 50°C, even when the instrument is turned off
  • The 135 mm diameter top is protected from chemicals and scratches by the superior resistant CeAlTop™ ceramic coating
POWERFUL STIRRING FOR ALL LAB APPLICATIONS
  • Brushless motor ensuring power and resistance over time
  • Stirs up to 20 L from 50 to 1500 rpm, making the Series suitable for both low and high-speed applications
  • SpeedServo™ counter reaction technology maintains the speed constant as viscosity changes
Specifications
Top Plate MaterialCeramic-coated Aluminum Alloy
Voltage230 V
Safety FeaturesOn Display (when the temperature is above 50°C)
Body MaterialEpoxy-painted Aluminum
Controller TypeDigital
Size135 mm dia.
Temperature (Metric)Ambient to 310°C
Temperature (English)Ambient to 590°F
Frequency50/60 Hz
Wattage630 W
Weight (English)3.75 lb.
Weight (Metric)1.7 kg
TypeStirring Hotplate
ShapeRound
Max. Sample Load20 L
Plug TypeEU/UK
No. of Positions1
Max. Container Size20 L
Stirring Range50 to 1500 rpm
Warranty3 Years
Dimensions (L x W x H)160 x 85 x 270 mm
DisplayDigital
Unit SizeEach