Analytical Balances
Actual product may vary
Analytical Balances
Featuring RS232 connectivity for easy communication, a backlit display and a simple interface for uncomplicated operation, these balances are perfectly designed for your workplace.
Have Questions?
Catalog NumberCapacityCalibration
FBS15927500220 gInternal
Catalog number FBS15927500
Price (SGD)
1,398.80
Special offer
Ends: 06-Sep-2026
2,152.00
Save 753.20 (35%)
Each
Capacity:
220 g
Calibration:
Internal
Price (SGD)
1,398.80
Special offer
Ends: 06-Sep-2026
2,152.00
Save 753.20 (35%)
Each
  • Large draftshield with sliding top and side doors
  • Standard RS-232 connectivity for easy data transfer
  • Stainless steel platform
  • Two adjustable feet that allow user to level the balance
  • Up-front level indicator - no need to look in back of balance during leveling process
  • Smart design and durable construction - small footprint which saves on benchtop space while providing a large weighing surface
  • Designed for uncomplicated operation with easy-to-use display and interface
  • Calibration lockout security loop in rear of balance
  • Weighing modes include weighing, parts counting and percent weighing
  • Includes AC adapter
  • Certifications: FCC, CE
  • Two year warranty
Specifications
CalibrationInternal
Certifications/ComplianceGLP with optional printer compliance
Connection TypeRS232
Capacity220 g
Display TypeBacklit LCD
Readability0.0001 g
IncludesAC adapter
Linearity0.0002 g
Min. Sample Weight200 mg
ModelFAS224
Repeatability0.0001 g
Stabilization Time4 sec.
TypeAnalytical Balance
Warranty2 Years
Weighing ModesWeighing, Parts Counting, Percent Weighing
Dimensions (D x W x H)317 x 201 x 303 mm
Diameter (Metric) Outer Pan90 mm
Electrical Requirements100/240 V ∼200 mA 50/60 Hz 12/18 VA
Amperage0.5 A