150 Homogenizer Package
150 Homogenizer Package
Actual product may vary
150 Homogenizer Package
Includes everything necessary to process samples as small as 200μL to 100mL. Fisherbrand™ 150 Homogenizer Package includes the 150w motor as well as accessories such as 2 saw tooth stainless steel generator probes, disposable generator probes and more.
Have Questions?
Catalog NumberPlug Type
FBO15595809UK Plug
Catalog number FBO15595809
Price (SGD)
0.00
EA
Add to cart
Plug Type:
UK Plug
Price (SGD)
0.00
EA
Add to cart
  • Processes laboratory samples, from 200μL to 100mL
  • Powerful 150w motor
  • Lightweight ergonomic design
  • Quiet operation, under 72db
  • Available with disposable generator probes (adapter for use is included)
  • Cross-contamination can be completely eliminated with the use of plastic generator probes
  • Stainless steel generator probes are recommended for larger and tougher samples
Specifications
Speed5000 to 35,000 rpm
Electrical Requirements220V UK Cord, 150W
Volume Range (Metric)200 μL to 100 mL
Diameter (Metric)5.1 cm
Height (Metric)16.5 cm
For Use With (Application)Tissue homogenization, tissue disruption, protein extraction, RNA extraction, DNA extraction, metabolite extraction, emulsification, proteomics, PCR, enzyme extraction, high speed stirring, powder suspension, genomics, metabolomics
Includes150 W motor, Flat bottom Stainless Steel generator probe (5 x 75 mm), Saw tooth Stainless Steel generator probe (7 x 115 mm), Saw tooth Stainless Steel generator probe (10 x 115 mm), Stand clamp, Motor stand assembly, Disposable probe adapter, Disposable generator probes, Instruction manual and Tool kit
Packaging Quantity1 Ea.
Plug TypeUK Plug
Weight (Metric)466 g
Unit SizeEA