150 and 850 Hand Held Homogenizer Accessory, Stainless Steel Probes
150 and 850 Hand Held Homogenizer Accessory, Stainless Steel Probes
Actual product may vary
150 and 850 Hand Held Homogenizer Accessory, Stainless Steel Probes
150 and 850 Hand Held Homogenizer Accessory, Stainless Steel Probes
150 and 850 Hand Held Homogenizer Accessory, Stainless Steel Probes
Choice of flat or saw tooth tips for use with hand-held homogenizers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBottom TypeVolume Range (Metric)Diameter (Metric) ProbeParticle Size
FBO15565819Sawtooth, wide window0.25 to 30 mL7 mmCourse
FBO15505829Sawtooth, wide window1.5 to 100 mL10 mmCourse
Catalog number FBO15565819
Price (SGD)
2,225.00
EA
Add to cart
Bottom Type:
Sawtooth, wide window
Volume Range (Metric):
0.25 to 30 mL
Diameter (Metric) Probe:
7 mm
Particle Size:
Course
Price (SGD)
2,225.00
EA
Add to cart
  • Provides end user flexibility to process samples
  • Designed for long life and are easy to disassemble for cleaning
  • Stator design allows fine to medium samples to pass through the processing windows
  • Comes with PTFE  upper and lower bearings
  • Autoclavable
Specifications
Product TypeProbe
Particle SizeCourse
Volume Range (Metric)0.25 to 30 mL
Diameter (Metric) Probe7 mm
Length (English) Probe4.5 in.
Length (Metric) Probe115 mm
MaterialStainless Steel
Bottom TypeSawtooth, wide window
For Use With (Equipment)150 Hand Held Homogenizer and 850 Homogenizers
Unit SizeEA