850 Homogenizer Accessory, Generator Probe
850 Homogenizer Accessory, Generator Probe
Actual product may vary
850 Homogenizer Accessory, Generator Probe
850 Homogenizer Accessory, Generator Probe
850 Homogenizer Accessory, Generator Probe
Stainless-steel generator probe; Fisher Scientific™ 850 Homogenizer Accessory, Generator Probe is compatible with Fisher Scientific 850 Homogenizers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberParticle SizeBottom TypeDiameter (Metric) ProbeLength (Metric) Probe
FBO15545829CoarseFlat, wide window30 mm195 mm
FBO15525829CoarseSawtooth, wide window20 mm195 mm
FBO15535829FineFlat30 mm195 mm
Catalog number FBO15545829
Price (SGD)
4,233.00
EA
Add to cart
Particle Size:
Coarse
Bottom Type:
Flat, wide window
Diameter (Metric) Probe:
30 mm
Length (Metric) Probe:
195 mm
Price (SGD)
4,233.00
EA
Add to cart
  • Comes with PTFE upper and lower bearings
  • Entire probe can be autoclaved
  • Easily disassembled for cleaning
Saw Tooth Generator Probe:
  • Add extra cutting action and stator design allows fine to medium samples to pass through processing windows
Specifications
Product TypeProbe
Particle SizeCoarse
Volume Range (Metric)200 mL to 5 L
Diameter (Metric) Probe30 mm
Length (Metric) Probe195 mm
Material316 Stainless Steel
Bottom TypeFlat, wide window
For Use With (Equipment)Fisher Scientific 850 Homogenizer
Unit SizeEA