Bead Mill 4 Mini Homogenizer
Actual product may vary
Bead Mill 4 Mini Homogenizer
Designed for personal use.
Have Questions?
Catalog NumberVoltage
FBO15525799115/230 VAC
Catalog number FBO15525799
Price (SGD)
4,800.00
Special offer
Ends: 06-Sep-2026
6,857.00
Save 2,057.00 (30%)
Each
Voltage:
115/230 VAC
Price (SGD)
4,800.00
Special offer
Ends: 06-Sep-2026
6,857.00
Save 2,057.00 (30%)
Each
  • Broad performance range, from gentle mixing to high force homogenization
  • Small footprint requires minimal bench space
  • Suited for homogenizing up to 4 samples in seconds
  • Reproducible extraction of proteins and nucleic acids
  • Simultaneously process 4 x 0.5 mL, 4 x 1.5mL, 4 x 2mL or 1 x 7mL samples
Specifications
Product TypeBead Mill Homogenizer
Voltage115/230 VAC
Description4-sample bead mill homogenizer
IncludesBead Mill 4 motor drive, integrated 0.5, 1.5 and 2mL tube carriage, 7mL tube carriage, tool kit, instruction manual
Unit SizeEach