850 Homogenizer Package
Actual product may vary
850 Homogenizer Package
Fisher Scientific™ 850 Homogenizer Package includes everything needed to disrupt, emulsify or homogenize a wide range of samples in volumes ranging from 200μL to 5L. Includes motor, stand assembly and 3 stainless steel probes.
Have Questions?
Catalog NumberVoltage
FBO15515819115 V
Catalog number FBO15515819
Price (SGD)
13,207.00
Special offer
Ends: 06-Sep-2026
18,867.00
Save 5,660.00 (30%)
Each
Voltage:
115 V
Price (SGD)
13,207.00
Special offer
Ends: 06-Sep-2026
18,867.00
Save 5,660.00 (30%)
Each
  • Includes 10 x 115mm, 20 x 195mm, and 30 x 195mm stainless steel generator probes
  • Powerful 850 watt motor
  • Integrated digital speed control displays speed and run time
  • Variable speed control from 5,000 to 25,000rpm
  • Can be used with plastic generator probes (adapter required for use)
  • Processing range from 0.2mL to 5L
  • Sound level: <72 db.
Specifications
Product TypeHomogenizer
Speed5000 to 25,000 rpm
Voltage115 V
Electrical Requirements115 V, 50/60 Hz
Height (English)8.5 in.
Height (Metric)21.5 cm
For Use With (Application)Tissue homogenization, tissue disruption, protein extraction, RNA extraction, DNA extraction, metabolite extraction, emulsification, proteomics, PCR, enzyme extraction, high speed stirring, powder suspension, genomics, metabolomics
Includes
  • 850 motor
  • 10x115 mm probe
  • 20x195 mm probe
  • 30x195 mm probe
  • 850 motor drive, tool kit, mounting stand clamp and instruction manual
  • Motor Stand
Weight (English)56 oz.
Weight (Metric)1.6 kg
Unit SizeEach