Check Order Status
Bulk Beads
Actual product may vary
Bulk Beads
For use with Bead Mill 4 and 24 Homogenizers
Have Questions?
Change view
Catalog Number
Particle Size
FBO15515809
1.4 mm
FBO15505809
2.38 mm
2 Options
Catalog number
FBO15515809
Price (SGD)
802.00
EA
Add to cart
Particle Size:
1.4 mm
Price (SGD)
802.00
EA
Add to cart
Fill up to 500 x 2mL sample tubes
Sample tubes not included
Specifications
Product Type
Beads
Particle Size
1.4 mm
Material
Zirconium Ceramic Oxide
For Use With (Equipment)
Bead Mill 4 and 24 Homogenizers
Packaging Quantity
325 g