Bead Mill 24 Homogenizer
Actual product may vary
Bead Mill 24 Homogenizer
Used for variety of applications that require grinding, lysing, and homogenization of biological samples prior to molecular extraction. Used with Fisherbrand™ Bead Kits.
Have Questions?
Catalog NumberDimensions (L x W x H)
FBO1551579917 x 15 x 13 in.
Catalog number FBO15515799
Price (SGD)
23,296.00
Each
Dimensions (L x W x H):
17 x 15 x 13 in.
Price (SGD)
23,296.00
Each
  • Simultaneously homogenize up to 24 x 2 mL or 0.5 mL tubes
  • More samples can be processed in smaller amount of time
  • Convenient front-loading tube holder position for ease of use and optimal bead/sample interactions
  • Includes 24 place tube carriage and Bead Mill Finger Plate that holds the 2mL tubes firmly in place during processing
  • Processing range is ideal for soft tissue samples
  • No cool-down required between runs (non-stop processing)
  • Disposable tubes ensure no threat of cross-contamination
  • No sample degradation due to quick processing
  • Smaller footprint to maximize bench space
  • Run Time: 1 to 99 sec.
  • Performance Range: 0.8 m/s to 6 m/s in increments of 1.5 m/s
  • Number of Cycles: 1-9
Specifications
Product TypeHomogenizer
Electrical Requirements110-230 VAC/50-60 Hz
Dimensions (D x W x H)17 x 15 x 13 in.
Dimensions (L x W x H)17 x 15 x 13 in.
For Use With (Application)DNA/RNA extraction, tissue homogenization, protein purification
Weight (English)65 lb.
Weight (Metric)29.5 kg
Unit SizeEach