Compact Digital Overhead Stirrer
Compact Digital Overhead Stirrer
Actual product may vary
Compact Digital Overhead Stirrer
Compact Digital Overhead Stirrer ideal for continuous, general purpose mixing. Easily fits into fume hoods or enclosed workstations. Includes chuck, chuck key, support rod, power cord, and instruction manual. Chuck is adjustable to 10 mm (3/8 in.). 220 Volt and includes two cord sets, UK and EU.
Have Questions?
Catalog NumberSpeed
FBFFSR2220U140 to 2002 rpm
Catalog number FBFFSR2220U1
Price (SGD)
2,768.00
EA
Add to cart
Speed:
40 to 2002 rpm
Price (SGD)
2,768.00
EA
Add to cart
  • Delivers accurate, repeatable results for volumes up to 25 L and 15,000 cps
  • Four-digit speed display
  • Three-button control (power on/off, speed increase, speed decrease, light on/off) is easy to use
  • Last speed setting is stored in memory before unit is powered off
  • Convenient handle makes stirrer easy to lift, move and set in place
  • Brushless, maintenance-free DC motor produces consistent mixing at all speeds and provides automatic overload protection
  • Belt-drive transmission is smooth, quiet and offers high power efficiency (1/15 hp, 50 w)
  • Sealed housing keeps liquids safely away from internal components
  • Through shaft allows quick, impeller adjustment and replacement
  • Includes support rod, chuck, chuck key, chuck guard, UK and EU cord sets, and printed manual
Specifications
Speed40 to 2002 rpm
Capacity25 L
DisplayDigital
Voltage220/240 V
Diameter (English) Shaft0.375 in.
Diameter (Metric) Shaft10 mm
Height (English)10 in.
Height (Metric)25.4 cm
Length (English)6 in.
Length (Metric)15.2 cm
Frequency50/60 Hz
Horsepower1/15 hp
Plug TypeEU/UK
Certifications/ComplianceCSA, CE, UL, FCC, IEC, ICES, EMC, EMI
IncludesOverhead stirrer motor, chuck, chuck key, support rod, external power supply, grounded power cord, manual in four languages (English, French, Spanish and German) and three year warranty
MaterialMolded Nylon and Cast Aluminum Housing
Motor TypeBrushless DC
Torque70 Ncm
TypeOverhead Stirrer
Weight (English)7 lb.
Weight (Metric)3.0 kg
Unit SizeEA