Propeller and Shaft
Propeller and Shaft
Actual product may vary
Propeller and Shaft
Propeller and Shaft
Propeller and Shaft
Pitched Blade Propeller and Shaft for use with Caframo™ and Fisherbrand™ Overhead Stirrers. This axial flow impeller is effective for high speed, low to medium viscosity mixing.
Have Questions?
Catalog NumberLength (Metric)
FBFA166400 mm
Catalog number FBFA166
Price (SGD)
420.00
EA
Add to cart
Length (Metric):
400 mm
Price (SGD)
420.00
EA
Add to cart
  • Electro-polished stainless steel 316L
  • Low shear, high top-to-bottom (axial) pumping capacity with high gas injection
  • Will vortex 5 gallons of water at 3600 rpm
  • High speed, Low to medium viscosity mixing, Axial flow pattern
Specifications
Diameter (English)2.5 in.
Diameter (English) Shaft0.3125 in.
Diameter (Metric)64 mm
Diameter (Metric) Shaft8 mm
Length (English)16 in.
Length (Metric)400 mm
ModelA166
TypePitched Blade Impeller
Weight (Metric)181 g
MaterialElectropolished Stainless Steel (316L)
Weight (English)0.4 lb.
Unit SizeEA