Clamp for Overhead Stirrer
Clamp for Overhead Stirrer
Actual product may vary
Clamp for Overhead Stirrer
Clamp for Overhead Stirrer
Clamp for Overhead Stirrer
Clamp for securing Caframo™ and Fisherbrand™ Overhead Stirrers.
Have Questions?
Catalog NumberDiameter (Metric)
FBFA12015 to 30 mm
Catalog number FBFA120
Price (SGD)
346.00
EA
Add to cart
Diameter (Metric):
15 to 30 mm
Price (SGD)
346.00
EA
Add to cart
  • Cast zinc-aluminum alloy coated with a chemical resistant epoxy paint
  • Clamps Overhead Stirrer to Stand
  • Includes place to hold chuck key
  • Weight: 640 g (1.5 lb.)
Specifications
Diameter (English)0.5625 to 1.1875 in.
Diameter (Metric)15 to 30 mm
For Use With (Equipment)Overhead Stirrer
TypeClamp
Weight (Metric)640 g
MaterialCast Zinc, Aluminum
Weight (English)1.5 lb.
Unit SizeEA