Sterile Polystyrene Disposable Serological Pipets with Magnifier Stripe
Actual product may vary
Sterile Polystyrene Disposable Serological Pipets with Magnifier Stripe
Sterile Polystyrene Disposable Serological Pipets with Magnifier Stripe
Sterile Polystyrene Disposable Serological Pipets with Magnifier Stripe
Corning
Sterile Polystyrene Disposable Serological Pipets with Magnifier Stripe
Pipets have improved optical clarity — high-contrast black markings make volumes easier to read
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Packaging TypeTolerance
FBC13-678-11E10 mLIndividual Paper/Plastic Wrap±0.2 mL
FBC13-678-11B1 mLIndividual Paper/Plastic Wrap±0.02 mL
FIS13-678-11B1 mLIndividual Paper/Plastic Wrap±0.02 mL
FIS13-676-10G1 mLIndividual All Plastic Wrap±0.02 mL
FKS118196601 mLIndividual All Plastic Wrap±0.02 mL
FBC13-678-11C2 mLIndividual Paper/Plastic Wrap±0.04 mL
FKS118591812 mLIndividual Paper/Plastic Wrap±0.04 mL
FIS13-675-3C2 mLIndividual All Plastic Wrap±0.04 mL
FBC13-678-11D5 mLIndividual Paper/Plastic Wrap±0.1 mL
FIS13-678-11D5 mLIndividual Paper/Plastic Wrap±0.1 mL
FKS118691815 mLIndividual Paper/Plastic Wrap±0.1 mL
FIS13-676-10H5 mLIndividual All Plastic Wrap±0.1 mL
FIS13-678-11E
also known as FIS#13-678-11E
10 mLIndividual Paper/Plastic Wrap±0.2 mL
FIS13-676-10J10 mLIndividual All Plastic Wrap±0.2 mL
FKS1183966010 mLIndividual All Plastic Wrap±0.2 mL
FIS13-676-10F10 mLBulk±0.02 mL
FKS1180966010 mLBulk±0.02 mL
FBC13-678-1125 mLIndividual Paper/Plastic Wrap±0.5 mL
FIS13-678-1125 mLIndividual Paper/Plastic Wrap±0.5 mL
FKS1183918125 mLIndividual Paper/Plastic Wrap±0.5 mL
FKS1151775225 mLIndividual All Plastic Wrap±0.5 mL
FBC13-678-11F50 mLIndividual Paper/Plastic Wrap±1 mL
FIS13-678-11F50 mLIndividual Paper/Plastic Wrap±1 mL
FKS1188918150 mLIndividual Paper/Plastic Wrap±1 mL
FIS13-676-10R50 mLIndividual All Plastic Wrap±1 mL
FKS1153775250 mLIndividual All Plastic Wrap±1 mL
FIS13-676-10Q50 mLBulk±1 mL
FKS1185966050 mLBulk±1 mL
FIS13-678-000100 mLIndividual Paper/Plastic Wrap±2%
FKS16348745100 mLIndividual Paper/Plastic Wrap±2%
Catalog number FBC13-678-11E
Price (SGD)
54.00
Case
Volume (Metric):
10 mL
Packaging Type:
Individual Paper/Plastic Wrap
Tolerance:
±0.2 mL
Price (SGD)
54.00
Case
The high-contrast black markings make the volumes easier to read, for improved optical clarity. Manufactured with optically clear polystyrene for accurate measurement. Fisherbrand™ Sterile Polystyrene Disposable Serological Pipets with Magnifier Stripe reduce reading errors and are designed for tissue culture work.
  • Color coded magnifier stripe runs lengthwise along pipet, making meniscus obvious at arm's length — reducing reading errors
  • Sterile and nonwettable pipets
  • Offer a tolerance of ±2% at full volume and have plug-in mouth end
  • 5, 10, 25, and 50mL pipets have ascending and descending graduations
  • Improved tip design on 25 and 50mL pipets increases the effect of surface tension to minimize dripping
  • Color coding on each pipet and on inner bags for multipack pipets make it easy to spot the size needed
  • Each pipet lot is tested for sterility, accuracy and endotoxin level
  • Bulk pack and individually wrapped, with paper and plastic wraps
  • Cotton plugged
Specifications
CalibrationTo Deliver/Blow Out
Certifications/ComplianceNon-pyrogenic, USP XXII, USP XXII Bacterial Endotoxins Test (85); DNase/RNase Free; BSE/TSE Free
MaterialPolystyrene
SterilitySterile
GraduatedYes
Graduations0.1 mL
Negative Graduations3 mL
Quantity200/Cs., 50/Pk., 4 Packs/Cs.
Tolerance±0.2 mL
Packaging TypeIndividual Paper/Plastic Wrap
ColorOrange
Volume (Metric)10 mL
TypeSerological Pipet