Check Order Status
Cardboard Cryogenic Freezer Box Partition/Divider
Actual product may vary
Cardboard Cryogenic Freezer Box Partition/Divider
Box for storage of small samples and tubes.
Have Questions?
Catalog Number
Holds
FBAHS2860H
81 Cryogenic Vials
Catalog number
FBAHS2860H
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Holds:
81 Cryogenic Vials
Array: 10 x 10
Low Temp: -196°C
Specifications
Color
White
Material
Cardboard
Shape
Square
Type
Box Divider
For Use With (Equipment)
1.5/2.0 mL Cryogenic Vials
Holds
81 Cryogenic Vials
Tube Array
9 x 9
Withstand Temperature (Metric)
-196°C