High Speed Mini-Centrifuge, 100/240 VAC, 50/60 Hz Universal Plug, Gray
High Speed Mini-Centrifuge, 100/240 VAC, 50/60 Hz Universal Plug, Gray
Actual product may vary
High Speed Mini-Centrifuge, 100/240 VAC, 50/60 Hz Universal Plug, Gray
High Speed Mini-Centrifuge, 100/240 VAC, 50/60 Hz Universal Plug, Gray
High Speed Mini-Centrifuge, 100/240 VAC, 50/60 Hz Universal Plug, Gray
Ideal for most protocols calling for fast spins and quick spindowns
Have Questions?
Catalog NumberCapacity
FBA12-006-90012 x 1.5/2 mL
Catalog number FBA12-006-900
Price (SGD)
1,015.00
Special offer
Ends: 31-Mar-2026
1,265.00
Save 250.00 (20%)
EA
Add to cart
Capacity:
12 x 1.5/2 mL
Price (SGD)
1,015.00
Special offer
Ends: 31-Mar-2026
1,265.00
Save 250.00 (20%)
EA
Add to cart
Ideal for most protocols calling for fast spins and quick spindowns
  • Compact footprint and brush-less motor for quiet operation and low vibration Select and set the digital speed display in RCF or rpm
  • Includes 12-place microtube rotor, 4 x 8 Strip 0.2 mL tube rotor and 0.2 mL and 0.5 mL tube adapters for the microtube rotor
  • Sample imbalance and tilt safety cut out
  • Safety interlock lid prevents opening while rotor is turning
  • Back lit display showing time, speed and operational information on the same screen
  • Quick-spin button for short spin requirements, press and hold to operate
  • Suitable for worldwide voltages 110 to 240 V, supplied with 4 different plug sets
  • UV-Resistant plastic, increased useful life under harsh light conditions
  • Double insulated, low voltage power supply
  • Push button lid release
  • Manual override allows user to open lid in case of power failure
Package includes:
  • High-Speed Mini-Centrifuge
  • Standard 12-Place, 1.5/2.0 mL Tube Rotor
  • 32-Place 0.2 mL Tube Rotor (singles, or 4 x 8 Strip)
  • 12 Tube Adapters, 0.2 mL
  • 12 Tube Adapters, 0.5 mL
Specifications
Capacity12 x 1.5/2 mL
Max. Speed12,500 rpm
Max. RCF2,000 x g
RefrigeratedNo
DisplayColor Graphic Display
Height (English) Exterior4.48 in.
Height (Metric) Exterior114 mm
Length (English) Exterior7.99 in.
Length (Metric) Exterior203 mm
Noise LevelLow
Width (English) Exterior6.73 in.
Width (Metric) Exterior171 mm
TypeMini-Centrifuge
IncludesMini-Centrifuge; 12-Place 1.5/2.0 mL Tube Rotor; 32-Place 0.2 mL Tube Rotor (singles, or 4 x 8 Strip); 12 Tube Adapters, 0.2 mL; 12 Tube Adapters, 0.5 mL
Electrical Requirements100/240 VAC, 50/60 Hz
Rotor IncludedYes
Dimensions (HWD)4.48 x 6.73 x 7.99 in. (11.4 x 17.1 x 20.3 cm)
Frequency50/60 Hz
Voltage100/240 VAC
Unit SizeEA