Thermolyne™ Benchtop Muffle Furnaces
Thermolyne™ Benchtop Muffle Furnaces
Actual product may vary
Thermolyne™ Benchtop Muffle Furnaces
Ideal for general laboratory use, including gravimetric analysis, sintering, quantitative analysis and heat treating.
Have Questions?
Catalog NumberCapacity (Metric)
F480585.8 L
Catalog number F48058
Price (SGD)
13,398.00
EA
Add to cart
Capacity (Metric):
5.8 L
Price (SGD)
13,398.00
EA
Add to cart
Ideal for general laboratory use, including gravimetric analysis, sintering, quantitative analysis and heat treating.
  • For added protection, the door safety switch stops power to heating elements when door opens
  • Thermocouple break protection cuts power to heating elements, preventing a thermocouple failure runaway condition
  • Two open coil heating elements on chamber sides assure fast heat-up with minimum temperature gradient
  • Thermal-efficient ceramic insulation surrounds chamber for maximum energy efficiency
  • 0.95cm (0.38 in.) dia. port for monitoring chamber temperatures with independent measuring device at rear of chamber
F47900 Models
  • 2L (0.07 cu. ft.) of heating area
F48000 Models
  • 5.9L (0.20 cu. ft.) of heating area
  • Includes ceramic shelf (SH480X1) that doubles furnace load capacity
A1 Controller
  • Digital single setpoint control
  • Single display shows actual temperature or setpoint
B1 Controller
  • Digital single setpoint control with a single ramp to setpoint and a dwell
  • Single display shows actual temperature or setpoint
  • Furnaces with this control also use a mechanical Overtemperature Protection relay
C1 Controller
  • Digital programmable control with one stored program of eight segments
  • Furnaces with this control also use a mechanical Overtemperature Protection relay
D1 Controller
  • Digital programmable control with four stored programs, 16 segments per program
  • With RS-232 communications interface
  • Furnaces with this control also use a mechanical Overtemperature Protection relay
Includes:
Power cord (-60 models include 20-amp cord; -33 units available with plug for China)
Warranty:
1 year (parts and labor)
Specifications
Capacity (Metric)5.8 L
Temperature (Metric)100°C to 1200°C
TypeSingle Setpoint, OTP
Shipping Weight (English)60 lb.
Unit SizeEA