▸
Ovens and Furnaces
▸
Furnaces
▸
Muffle Furnaces
▸
Thermolyne™ Benchtop Muffle Furnaces
Actual product may vary
Thermo Scientific
Thermolyne™ Benchtop Muffle Furnaces
Compact, lightweight benchtop furnaces
Have Questions?
Catalog Number
Capacity (Metric)
Voltage
Controller Type
F48010
5.8 L
240 V
Digital single setpoint
Catalog number
F48010
HSN
85141900
Price (SGD)
3,914.00
Special offer
Ends:
30-Jun-2026
4,892.00
Save 978.00 (20%)
Each
Add to cart
Capacity (Metric):
5.8 L
Voltage:
240 V
Controller Type:
Digital single setpoint
Price (SGD)
3,914.00
Special offer
Ends:
30-Jun-2026
4,892.00
Save 978.00 (20%)
Each
Add to cart
Compact, lightweight benchtop furnaces
For added protection, the door safety switch stops power to heating elements when door opens
Thermocouple break protection cuts power to heating elements, preventing a thermocouple failure runaway condition
Two open coil heating elements on chamber sides assure fast heat-up with minimum temperature gradient
Thermal-efficient ceramic insulation surrounds chamber for maximum energy efficiency
0.95cm (0.38 in.) dia. port for monitoring chamber temperatures with independent measuring device at rear of chamber
F47900 Models
2L (0.07 cu. ft.) of heating area
F48000 Models
5.9L (0.20 cu. ft.) of heating area
Includes ceramic shelf (SH480X1) that doubles furnace load capacity
A1 Controller
Digital single setpoint control
Single display shows actual temperature or setpoint
B1 Controller
Digital single setpoint control with a single ramp to setpoint and a dwell
Single display shows actual temperature or setpoint
Furnaces with this control also use a mechanical Overtemperature Protection relay
C1 Controller
Digital programmable control with one stored program of eight segments
Furnaces with this control also use a mechanical Overtemperature Protection relay
D1 Controller
Digital programmable control with four stored programs, 16 segments per program
With RS-232 communications interface
Furnaces with this control also use a mechanical Overtemperature Protection relay
Specifications
Controller Type
Digital single setpoint
Capacity (Metric)
5.8 L
Temperature (Metric)
100°C to 1200°C
Type
Muffle Furnace
Voltage
240 V
Wattage
1800 w
Amperage
7.5 A
Frequency
50/60 Hz