Check Order Status
Scratch Brush
Actual product may vary
Scratch Brush
This Scratch Brush is excellent for presolder and contact cleaning.
Have Questions?
Catalog Number
Product Type
Size
Bristle Material
ECT267
Scratch Brush
0.375 x 4.75 in.
Fiberglass
Catalog number
ECT267
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Product Type:
Scratch Brush
Size:
0.375 x 4.75 in.
Bristle Material:
Fiberglass
Fine precision:
Retractable and refillable
With plastic handles
Fine deburring:
Polishing
Rust removal
Cleaning and erasing in chemical etching processes
Specifications
Product Type
Scratch Brush
Bristle Material
Fiberglass
Color-coded
No
Handle Material
Plastic
Diameter (English) Bristle
0.375 in.
Diameter (Metric) Bristle
0.96 cm
For Use With (Application)
Fine deburring, polishing, rust removal, cleaning and erasing in chemical etching processes
Length (English)
4.75 in.
Length (Metric)
12 cm
Precision
Fine
Size
0.375 x 4.75 in.