Benchtop Coolers
Benchtop Coolers
Actual product may vary
Benchtop Coolers
Benchtop Coolers
Benchtop Coolers
Benchtop Coolers
Benchtop Coolers
Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature RangeArrayHolds
DS5116-00320°C4 x 832 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
FIS09-74072-20°C4 x 520 Tubes (0.2 to 0.5 mL)
FKS10777232-20°C4 x 520 Tubes (0.2 to 0.5 mL)
5115-0012-20°C3 x 412 Tubes (0.2 to 0.5 mL)
FIS15-350-61-20°C3 x 412 Tubes (0.2 to 0.5 mL)
5115-0032-20°C4 x 832 Tubes (0.2 to 0.5 mL)
5116-13000°C3 x 412 Tubes (12 to 13 mm dia.)
FIS15-350-630°C3 x 412 Tubes (12 to 13 mm dia.)
FIS15-350-530°C3 x 412 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
FIS15-350-540°C4 x 832 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
FIS15-350-62-20°C3 x 412 Cryogenic Vials (0.2 to 1.5 mL)
FKS10741792-20°C3 x 412 Cryogenic Vials (0.2 to 1.5 mL)
Catalog number DS5116-0032
Price (SGD)
551.00
CS
Add to cart
Temperature Range:
0°C
Array:
4 x 8
Holds:
32 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
Price (SGD)
551.00
CS
Add to cart
Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.
  • Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
  • Holds 12, 20 or 32 tubes
  • Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
  • Printed grid on one-way keyed lid for proper orientation and sample identification
  • Molded of impact-resistant polycarbonate with locking handle for long-term use
  • Stackable to save space
Specifications
Height (Metric)146 mm
Height (English)5.75 in.
Length (Metric)243 mm
Length (English)9.56 in.
Width (Metric)157 mm
Width (English)6.19 in.
Dimensions (L x W x H)9.56 x 6.19 x 5.75 in. (243 x 157 x 146 mm)
Description4x8 compartments, gel filled, 0 deg.
Array4 x 8
Holds Temp., Hours<1°C, 5.0
DesignGel-filled/white lid
MaterialPolycarbonate
TypeBenchtop Cooler
ColorBlue, Clear
Capacity (Metric)0.2 to 0.5 mL
Holds32 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
Temperature Range0°C
Unit SizeCS