Nalgene™ PFA Graduated Plastic Beakers with Handles
Nalgene™ PFA Graduated Plastic Beakers with Handles
Actual product may vary
Nalgene™ PFA Graduated Plastic Beakers with Handles
Nalgene™ PFA Graduated Plastic Beakers with Handles
Nalgene™ PFA Graduated Plastic Beakers with Handles
Nalgene™ PFA Graduated Plastic Beakers with Handles
Conduct high-purity applications that use strong oxidizing agents, acids, hydrocarbons and ketones with these PFA graduated beakers with handles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
DS1511-30003000 mL
FIS02-592-5B3000 mL
Catalog number DS1511-3000
Price (SGD)
718.00
CS
Add to cart
Capacity (Metric):
3000 mL
Price (SGD)
718.00
CS
Add to cart
Conduct high-purity applications that use strong oxidizing agents, acids, hydrocarbons and ketones with Thermo Scientific™ Nalgene™ PFA Graduated Beakers with Handles. These beakers feature extraordinary chemical resistance. PFA is non-stick, making the beaker easy to clean and resistant to contamination.
  • Withstands exposure to extreme temperatures from -270° to +250°C
  • Rigid and translucent, enabling easy and consistent viewing of liquid levels
  • Molded-in graduation in both oz. and mL won't wear off with continuous use or washings
  • Excellent chemical resistance to strong ozidizing agents, acids, hydrocarbons and ketones make these beakers useful for a broad range of high-purity applications
  • Molded-in rectangular handle for easy, safe handling and pouring
  • Autoclavable
.
Specifications
ColorNatural translucent
DescriptionBeaker with Handle PFA 3000 mL
Capacity (Metric)3000 mL
MaterialPFA
TypeBeaker
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
ScaleMolded Dual Scale
StyleHandled
Unit SizeCS