Human Papillomavirus (HPV) Verification Panel
Human Papillomavirus (HPV) Verification Panel
Actual product may vary
Human Papillomavirus (HPV) Verification Panel
Microbiologics™ Human Papillomavirus (HPV) Verification Panel includes 20 inactivated microorganism pellets each comprised of the 3 organisms: Recombinant HPV 16, Recombinant HPV 18, and Recombinant HPV 45
Have Questions?
Catalog NumberQuantity
DLB821620
Catalog number DLB8216
Price (SGD)
-
Quantity:
20
  • Pellets are stable at room-temperature
  • For in vitro diagnostic use
  • Control includes DNA sequences for E6/E7 amplification of HPV 16, 18 and 45 types
  • Does not contain human beta globin
  • Currently not available for shipment to Canada
Specifications
Certifications/ComplianceIVD, CE Marked
FormatHelix Elite
Manufacturing OriginUnited States
Product TypeHelix Elite Control Reference Panels and Standards
For Use With (Application)In vitro diagnostics
Packaging TypePlastic Box
Quantity20
Shelf Life8 Months (Minimum)
Unit SizeCase
Contents & Storage
Room Temperature