Barnstead™ E-Pure™ Ultrapure Water Purification Systems
Barnstead™ E-Pure™ Ultrapure Water Purification Systems
Actual product may vary
Barnstead™ E-Pure™ Ultrapure Water Purification Systems
Produce Type 1 ultrapure water with low operating costs using the Thermo Scientific™ Barnstead™ E-Pure™ Ultrapure Water Purification system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModulesVoltage
D4642-334240 V
FKS114663484240 V
Catalog number D4642-33
Price (SGD)
11,163.00
EA
Add to cart
Modules:
4
Voltage:
240 V
Price (SGD)
11,163.00
EA
Add to cart
Produce Type 1 ultrapure water with low operating costs using Thermo Scientific™ Barnstead™ E-Pure™ Ultrapure Water Purification Systems. Choose from a three- or four-module system. Gain longer cartridge life and benefit from lower operating costs with large cartridges. This system is Ideal for AA, GS, buffer preps, or as a purified feed water source for analytical instruments or labware washers. It is simple to use and easy to maintain.
AquaPro™ digital on-line analytical measurement platform featuring:
  • Large color graphic display
  • Temperature readout
  • Temperature compensated
  • Calibratable
Features:
  • System achieves resistivities up to 18.2 MΩ-cm and TOC levels below 10ppb.
  • Virgin polypropylene water pathways prevent recontamination.
  • Recirculation pump continuously recirculates water throughout the entire system to maintain water purity and reduce the quantity of wastewater required for rinse-up.
  • System is wall-mountable and completely assembled for easy installation.
  • Quarter-turn quick release canisters allow for easy cartridge replacement.
  • 0.2μm absolute final filter removes bacteria and particulates that could alter your results.
  • Digital resistivity meter is automatically temperature-compensated to 25°C, preventing fluctuating readings due to temperature changes.
Ordering Information:
Optional accessories include Remote Dispenser (D8952), Low Pressure Pump Protector (D2706), and Low Water Level Pump Protector (D0603, D0606). These products, as well as replacement cartridges and consumables, are available separately. For details, contact your sales representative.
Includes:
Wall bracket
Requires:
O-rings included with cartridge pack (sold separately)
Recommended for:
AA, GS, buffer preps, feedwater for analytical instruments or labware washers
Note:
If feeding E-Pure from a storage reservoir, Low Water Pump Protector is recommended to protect pump from damage if reservoir water level falls below system requirements. Order separately (D0603 for 120V, D0606 for 240V).
Specifications
Certifications/ComplianceCE, CSA
Conductivity0.055μS/cm
Bacterial Content<1 CFU/mL
Feed Water MonitoringNo
Feedwater SourceTap or Pretreated
Flow RateUp to 2.5 L/min.
Product Water TypeType 1
Remote DispenserOptional
Resistivity18.2 Mohm.cm at 25°C
TOC Monitor with UV Intensity MonitoringNo
Total Organic Carbon<10 ppb
TypeWater Purification System
Feedwater Pressure100 psig (6.9 bar)
Modules4
Particles >0.2µM/mL<1 μM/mL
Dimensions (L x W x H)7.5 x 37.5 x 28 in. (19 x 95.3 x 71.1 cm)
Length (English)7.5 in.
Length (Metric)19 cm
Width (English)37.5 in.
Width (Metric)95.3 cm
Height (English)28 in.
Height (Metric)71.1 cm
Voltage240 V
Unit SizeEA