B-Pure™ Water Purification System
B-Pure™ Water Purification System
Actual product may vary
B-Pure™ Water Purification System
Customize the economical Thermo Scientific™ B-Pure™ laboratory-grade water system to meet your pretreatment or deionization application needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
D4521Double Holder Module with Digital Purity Meter
D4511Water Purification Single Holder Module
Catalog number D4521
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
Double Holder Module with Digital Purity Meter
Customize the economical Thermo Scientific™ B-Pure™ laboratory-grade water system to meet your pretreatment or deionization application needs. The modular design provides the ability to add additional holders when the need arises. Choose from single, double or half-size holders. Easily change cartridges with the simple quarter turn quick release canisters.
Features:
  • Produces up to 4L/min., depending on model
  • Quarter-turn quick-release canisters make cartridge changes easy
  • Accepts Thermo Scientific Pura-Lite™ indicator or digital purity meter to monitor water quality
Single Module:
  • Simple system without draw-off valve or purity indicator
  • Produces up to 4L/min.
Double Module:
  • Complete with draw-off valve
  • Produces up to 4L/min.
  • Choose from models with digital purity meter or Pura-Lite “Go/No Go” resistivity indicator
Half-Size Holder:
  • Produces up to 2L/min.
  • Easily attaches to full-size B-Pure systems
  • Small size for flexibility in mounting location
Requires:
Cartridges (sold separately).
Recommended for:
Pretreatment, deionization
Note:
The Thermo Scientific H2O Select Program is available to help select the correct B-Pure system. For details, contact your sales representative.
Specifications
Certifications/ComplianceCSA
DescriptionDouble holder module with digital purity meter; 120VAC
Flow Rate4 L/min.
Feedwater Temperature (English)40°F to 120°F
Feedwater Temperature (Metric)4°C to 49°C
For Use With (Equipment)Water Purification Double Holder Module
Inlet Connection NPTF (English)0.5 in.
Inlet Connection NPTF (Metric)1.3 cm
Inlet Pressure100 psig
Requires2 Full-Size B-Pure Cartridges
Resistivity1 to 10MΩ·cm (depending on cartridge used)
TypeDouble Holder Module with Digital Purity Meter
Dimensions (L x W x H)7 x 15 x 27 in. (17.8 x 38.1 x 68.6 cm)
Length (English)7 in.
Length (Metric)17.8 cm
Width (English)15 in.
Width (Metric)38.1 cm
Height (English)27 in.
Height (Metric)68.6 cm
Voltage120 V
Unit SizeEA