Barnstead™ Final Filter
Barnstead™ Final Filter
Actual product may vary
Barnstead™ Final Filter
Use the Thermo Scientific™ Barnstead™ Final Filter on select models of Barnstead pure and ultrapure water systems.
Have Questions?
Catalog NumberPore Size
D37500.2 μm
Catalog number D3750
Price (SGD)
280.00
EA
Add to cart
Pore Size:
0.2 μm
Price (SGD)
280.00
EA
Add to cart
Use the Thermo Scientific™ Barnstead™ Final Filter on select models of Barnstead pure and ultrapure water systems. Compatible with Thermo Scientific™ B-Pure™, Nanopure™, Easypure™ II, Easypure™ II RF, Easypure™ RoDi, E-Pure™ (3,4 holder) and TII systems.
Specifications
Description0.2µm Final Filter
Pore Size0.2 μm
For Use With (Equipment)B-Pure, Nanopure, Easypure II, Easypure II RF, Easypure RoDi, E-Pure (3,4 holder), and TII systems
TypeFinal Filter
Unit SizeEA