Owl™ D2 Wide-Gel System Replacement Parts
Owl™ D2 Wide-Gel System Replacement Parts
Actual product may vary
Owl™ D2 Wide-Gel System Replacement Parts
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
Have Questions?
Catalog NumberType
D2-UVTGel Tray
Catalog number D2-UVT
Price (SGD)
516.00
EA
Add to cart
Type:
Gel Tray
Price (SGD)
516.00
EA
Add to cart
Replace the UVT gel tray, gaskets and external caster with Thermo Scientific™ Owl™ D2 Wide-Gel System Replacement Parts, for use with Owl™ D2 Wide-Gel System.
  • UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
  • Comb slots are positioned so you can arrange up to 12 sample sets on the same gel
Specifications
DescriptionEasyCast™ gasketed UVT gel tray
For Use With (Equipment)Owl™ D2 Wide Gel Electrophoresis System
Product LineOwl
TypeGel Tray
Length (Metric)10.2 cm
Width (Metric)14.4 cm
Unit SizeEA