Deionizer Cartridges
Deionizer Cartridges
Actual product may vary
Deionizer Cartridges
Deionizer Cartridges
Deionizer Cartridges
Choose from an array of Thermo Scientific™ Deionizer Cartridges for select Thermo Scientific™ Barnstead™ water systems, based on your application.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Resin TypeCapacityFor Use With (Application)
D0832Bantam™ Deionizer, B-Pure™Mixed Bed, Activated Carbon785 grains/1000 gal.Ultrapure and Organic Removal
D0760Bantam™ Deionizer, B-Pure™Strong Anion Exchange1680 grainsAnion Removal
FIS09-034-92Bantam™ Deionizer, B-Pure™Strong Anion Exchange1680 grainsAnion Removal
D0809Bantam™ Deionizer, B-Pure™Mixed Bed875 grains
FIS09-034-3Bantam™ Deionizer, B-Pure™Mixed Bed875 grains
D0811B-Pure™Porous Strong Ion Exchange30 gOxygen Removal
D0813Bantam™ Deionizer, B-Pure™Activated Carbon5000 gal.Organic Removal
FIS09-034-5Bantam™ Deionizer, B-Pure™Activated Carbon5000 gal.Organic Removal
D0815Bantam™ Deionizer, B-Pure™Strong Cation Exchange3170 grainsCation Removal
FIS09-034-10Bantam™ Deionizer, B-Pure™Strong Cation Exchange3170 grainsCation Removal
FIS09-028-33Bantam™ Deionizer, B-Pure™Mixed Bed, Activated Carbon785 grains/1000 gal.Ultrapure and Organic Removal
D0836B-Pure™Macroreticular, Activated Carbon2000 gal.Pretreatment Macropure
Catalog number D0832
Price (SGD)
286.00
EA
Add to cart
For Use With (Equipment):
Bantam™ Deionizer, B-Pure™
Resin Type:
Mixed Bed, Activated Carbon
Capacity:
785 grains/1000 gal.
For Use With (Application):
Ultrapure and Organic Removal
Price (SGD)
286.00
EA
Add to cart
Choose from an array of Thermo Scientific™ Deionizer Cartridges based on your application needs. These cartridges are for use with the Thermo Scientific™ Barnstead™ Bantam™, B-Pure™ or Nanopure™ water purification systems as indicated.
  • Choice of full or half cartridges
Specifications
DescriptionCartridge for ultrapure and organic removal; mixed-bed, activated carbon; full size
Capacity785 grains/1000 gal.
Resin TypeMixed Bed, Activated Carbon
For Use With (Application)Ultrapure and Organic Removal
For Use With (Equipment)Bantam™ Deionizer, B-Pure™
TypeDeionizer Cartridge
Unit SizeEA