Bantam™ Deionizer
Bantam™ Deionizer
Actual product may vary
Thermo Scientific
Bantam™ Deionizer
Customize this economical water deionizer to a specific application with a variety of cartridges. Resistivity is easily viewed directly from the unit.
Have Questions?
Catalog NumberVoltage
D0805240 V
Catalog number D0805
Price (SGD)
3,212.00
EA
Add to cart
Voltage:
240 V
Price (SGD)
3,212.00
EA
Add to cart
Customize the Thermo Scientific™ Bantam™ Deionizer to your specific application by selecting the appropriate cartridge. An ideal choice for an economical way to deionize water, this deionizer allows you to easily view resistivity directly from the unit.
Features:
  • Ideal for applications requiring up to 38L/hr. of purified water
  • Point-of-use polishing of pretreated water or single-stage treatment of tap water
  • Direct-reading purity monitor
  • Mounts on benchtop or wall
Includes:
  • 3′ (91cm) inlet tubing
Requires:
Recommended for:
  • Pretreatment
  • Deionization
Note:
Designed for nonpressurized applications. Output water must be diverted to an atmospherically vented receptacle; no back pressure can be accepted.
Specifications
DescriptionBantam Deionizer, 240V
Flow RateUp to 38 L/hr.
Inlet Pressure5 to 70 psi
ModelBantam Deionizer
Resistivity25,000 to 18,000,000Ω·cm
TypeDeionizer
Dimensions (L x W x H)8.75 x 6 x 28.5 in. (22 x 15 x 72 cm)
Length (English)8.75 in.
Length (Metric)22 cm
Width (English)6 in.
Width (Metric)15 cm
Height (English)28.5 in.
Height (Metric)72 cm
Electrical Requirements240 V 50/60 Hz
Voltage240 V