Replacement Probe
Product Image
Replacement Probe
Use this Fast-response RTD platinum probe with thin micro cable for high-precision results. Traceable™ Replacement Probe is designed for use with Traceable -100.0 Platinum Freezer Thermometer.
Have Questions?
Catalog NumberProbe Type
CTL4231RTD Platinum Probe
Catalog number CTL4231
Price (SGD)
-
Probe Type:
RTD Platinum Probe
Use this Fast-response RTD platinum probe with thin micro cable for high-precision results. Traceable™ Replacement Probe is designed for use with Traceable -100.0 Platinum Freezer Thermometer.
  • For use with digital thermometer no. 4230
Specifications
Probe TypeRTD Platinum Probe
Product TypeReplacement Probe
For Use With (Equipment)Traceable 100.0 Platinum Freezer Thermometer
Unit SizeEA