Traceable™ Memory-Loc™ Datalogging Thermometer
Actual product may vary
Traceable™ Memory-Loc™ Datalogging Thermometer
Traceable™ Memory-Loc™ Datalogging Thermometer that can be used in 21 CFR 11 environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
CTL15-081-1321 Probe, 5/16 in. dia. x 3/4 in. long
CTL15-081-1342 Probes (Bottle Probe and 5/16 in. dia. x 3/4 in. long)
CTL15-081-1301 Bottle
CTL15-081-1362 Stainless-steel Probes
CTL15-081-1312 Bottles
CTL15-081-1351 Stainless-steel Probe
CTL15-081-1332 Probes, 5/16 in. dia. x 3/4 in. long
Catalog number CTL15-081-132
Price (SGD)
581.00
Each
Includes:
1 Probe, 5/16 in. dia. x 3/4 in. long
Price (SGD)
581.00
Each
Traceable™ Memory-Loc™ Datalogging Thermometer that can be used in 21 CFR 11 environments.
  • Data-logging thermometer with fixed one-minute timing intervals
  • Meets current CDC requirements and additional recommendations for vaccine storage and monitoring
  • Can be used in 21 CFR 11 environments, raw data is locked and cannot be cleared or changed on base unit
  • Stores up to ten unique alarm events
  • Capable of storing over 1 million temperature observations
  • Hassle-free retrieval of data; unit can remain in use while downloading and analyzing data
  • Status indicators: Low battery, low memory, USB data transfer and active alarm state
  • Traceable to NIST for accuracy
  • High-impact, chemical-resistant ABS plastic case measures 7 x 10.8 x 1.9 cm (2.75 x 4.25 x 0.75 in.)
  • Weight: 5 oz.
Hi/Lo Alarms and Time/Date Stamps
  • Alarm tracking feature stores in memory each alarm event detailing when the alarm state occurred, as well as when the unit returned back to within range
  • Alarm state indicators include visual LEDs, audio alerts, and flashing LCD segments
  • Data for up to ten different alarm events can be retrieved with the most current event viewable on the display
USB Data Transferring
  • All new feature allows recorded data (CSV file) to be transferred from thermometer to PC (or Mac™) using a USB flash drive (not included)
  • No software is needed for computer interface
  • Each thermometer has a unique ID allowing multiple units to be used in the same location
  • Raw data on unit cannot be overwritten or modified
  • Indicator light will appear when 95% of memory is full
Data-Logging Features
  • Can be used in environments requiring 21 CFR 11 compliance
  • Thermometer logs two years of observations with a one-minute logging interval
  • Device will stop recording when storage is full and will not overwrite recorded data
  • Data is unable to be deleted from memory once stored
Bottle Probe Features
  • Bottle probes are sealed in a miniature bottle (1 x 2.5 in.) filled with nontoxic glycol
  • Solution is GRAS (generally recognized as safe) by the FDA (Food and Drug Administration)
  • Eliminates concerns about incidental contact with food or drinking water
  • Temperature-buffered bottle sensor eliminates rapid changes when refrigerator door is opened
  • Ideal for use in monitoring storage conditions for vaccines or other important samples
  • Meets all current CDC requirements and recommendations for temperature monitoring of vaccine storage as part of the Vaccines for Children program
Specifications
TypeDatalogger
Display TypeLCD
CalibrationNot Calibrated
Alarm TypesLED Indicator, Audible, Flashing LCD Segment
Software TypeTraceableLIVE
Description1 Probe, 5/16 in. dia. x 3/4 in. long
Includes1 Probe, 5/16 in. dia. x 3/4 in. long
Certifications/ComplianceNIST, ISO/IEC 17025, A2LA, CDC
InterfaceUSB
Sensor TypesTemperature-Buffered Bottle Sensor
Accuracy (Temperature)±0.25°C
Temperature Resolution0.01°
Temperature Range (Metric)-50°C to +70°C
Operating Temperature (English)-58°F to +158°F
Operating Temperature (Metric)-72.4°F to +316.4°F
Weight (English)5 oz.
Weight (Metric)141.7 g
Unit SizeEach