Traceable™ 100-Hour Four-Channel Timer
Actual product may vary
Traceable™ 100-Hour Four-Channel Timer
Fisherbrand™ Traceable™ 100-Hour Four-Channel Timer is a lab workhorse provides 100 hour timing on four channels.
Have Questions?
Catalog NumberTime Range
CTL15-079-69799 hr. 59 min. and 59 sec. on 4 channels
Catalog number CTL15-079-697
Price (SGD)
93.00
Each
Time Range:
99 hr. 59 min. and 59 sec. on 4 channels
Price (SGD)
93.00
Each
Fisherbrand™ Traceable™ 100-Hour Four-Channel Timer is a lab workhorse provides 100 hour timing on four channels.
  • Alarm sounds for 60 seconds and may be silenced manually
  • Includes count/down feature and time-of-day clock
Specifications
Product TypeTimer
Time Range99 hr. 59 min. and 59 sec. on 4 channels
AlarmsAudible
Accuracy0.01%
DisplayDigital
DescriptionUSB Interface
No. of Channels4
Battery TypeAAA
Length (Metric)10.16 cm
Weight (English)1.5 oz.
Weight (Metric)0.04 kg
IncludesBattery, Flip-Open Stand, Traceable™ Certificate
Unit SizeEach