Traceable™ Digital Thermometer with Stainless-Steel Stem, 0.25 in. LCD Screen, and Protective Guard
Traceable™ Digital Thermometer with Stainless-Steel Stem, 0.25 in. LCD Screen, and Protective Guard
Actual product may vary
Traceable™ Digital Thermometer with Stainless-Steel Stem, 0.25 in. LCD Screen, and Protective Guard
Easy-to-read handheld Traceable™ Digital Thermometer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTemperature Range (Metric)Temperature Range (English)AccuracyLength (Metric) Probe
CTL15-078G-50°C to +300°C-58°F to +572°F±1.5°C8.9 cm
CTL06-664-33-50°C to +300°C-58°F to +572°F±0.4°C8.9 cm
CTL06-664-32-50°C to +280°C-58°F to +536°F±0.4°C20 cm
CTL14-648-12-50°C to +150°C-58°F to +302°F±0.2°C20 cm
CTL15-077-54-50°C to +280°C-58°F to +536°F±1°C20.32 cm
CTL15-077-59-50°C to +250°C-58°F to +482°F±1°C28.87 cm
CTL15-077-61-50°C to +300°C-58°F to +572°F±0.5°C28.87 cm
CTL15-078-G-50°C to +300°C-58°F to +572°F±1.5°C8.9 cm
CTL15-078-J-50°C to +150°C-58°F to +302°F±1°C (-20°C to +120°C)20.32 cm
Catalog number CTL15-078G
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Temperature Range (Metric):
-50°C to +300°C
Temperature Range (English):
-58°F to +572°F
Accuracy:
±1.5°C
Length (Metric) Probe:
8.9 cm
Easy-to-read handheld Traceable™ Digital Thermometer.
  • Longer probes are excellent for measuring sample temperatures while stirring in deep vessels or water baths
  • Thermometers with longer sampling rates allow thermometer to be removed from sample before reading—ideal when display is difficult to read in measuring position
  • For use in cuvets, soil, incubators, petroleum products and foods
Specifications
Certifications/ComplianceISO 17025, A2LA, NIST
Diameter (English) Probe0.14 in.
Diameter (Metric) Probe3.56 mm
For Use With (Application)Measuring temperature in soil, cuvettes, test tubes, food products, incubators and petroleum hydrocarbons
IncludesTraceable certificate, Protective case
Length (English) Probe3.5 in.
Length (Metric)177 mm
Length (Metric) Probe8.9 cm
MaterialPlastic
Resolution0.1°C
Weight (English)1 oz.
Weight (Metric)3175 g
Product TypeDigital Thermometer
Length (English)7 in.
Max. Temperature (English)572°F
Max. Temperature (Metric)300°C
Min. Temperature (English)-58°F
Min. Temperature (Metric)-50°C
Temperature Range (English)-58°F to +572°F
Temperature Range (Metric)-50°C to +300°C
CalibratedYes
Accuracy±1.5°C
DisplayDigital LCD
Width (Metric)8 cm
Unit SizeEA