Traceable™ Infrared Thermometer with Pocket Clip
Actual product may vary
Traceable™ Infrared Thermometer with Pocket Clip
Pocket IR thermometer with simple point-and-shoot operation
Have Questions?
Catalog NumberTemperature Range (Metric)
CTL15-077-966-33°C to +220°C
Catalog number CTL15-077-966
Price (SGD)
165.00
Each
Temperature Range (Metric):
-33°C to +220°C
Price (SGD)
165.00
Each
Easy to use — just turn it on and take a reading in less than a second. Fisher Scientific™ Traceable™ Infrared Thermometer with Pocket Clip offers simple point-and-shoot operation.
  • Units read both Fahrenheit and Celsius of any surface—solids, semisolids, and liquids
  • Noninvasive, no-touch measurements are ideal for food preparation, life sciences, pharmaceuticals, petroleum products, cleanrooms, electronics, and field use
  • 'HOLD' command freezes the display at the current reading Field of view is 1:1
  • Display indicates low battery
  • Fixed emissivity of 0.95 covers 90% of all applications
Specifications
  • Resolution: 0.1°
  • Accuracy: ±1°C + 2%
  • Dimensions: 3.25 x 0.75 in. (8.3 x 1.9cm)
  • Weight: 0.75 oz.
Specifications
Certifications/ComplianceISO 17025, NIST
Emissivity0.95 Fixed
For Use With (Application)Food, life sciences, pharmaceuticals, petroleum products, clean rooms, electronics and field
IncludesHandy pocket clip, key chain strap, battery
Resolution0.1°C
Weight (English)0.75 oz.
Weight (Metric)0.021 kg
Product TypePocket Infrared Thermometer
Max. Temperature (English)428°F
Max. Temperature (Metric)220°C
Min. Temperature (English)-27.4°F
Min. Temperature (Metric)-33°C
Temperature Range (Metric)-33°C to +220°C
CalibratedYes
Battery TypeLR44 1.5V
Accuracy±1°C
DisplayDigital
Width (Metric)1.9 cm
Unit SizeEach