Logger-Trac™ Mini-Logger-Trac Datalogging Thermometer
Logger-Trac™ Mini-Logger-Trac Datalogging Thermometer
Actual product may vary
Logger-Trac™ Mini-Logger-Trac Datalogging Thermometer
Know the temperature excursion duration immediately, no data download required with Logger-Trac™ Mini-Logger-Trac Datalogging Thermometer.
Have Questions?
Catalog number CTL14-650-122
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Know the temperature excursion duration immediately, no data download required with Logger-Trac™ Mini-Logger-Trac Datalogging Thermometer.
  • Duration of Temperature excursion
  • Data-logger with user-defined recording intervals
  • Active LCD Display shows the following important data: Current temperature, Minimum/Maximum temperatures, Recording status, Run Time, Alarm (Excursion) Cumulative Time, Alarm Tally
  • Requires the Fisherbrand Traceable Cradle and USB Cable (6549) to upload data to PC
USER SELECTABLE RECORDING INTERVALS For complete flexibility, the user-defined recording interval may be set based on the number of desired days to log (from 4 hours to 365 days) or the desired time interval (from every 2 seconds to every 24 hours).Internal flash memory stores 7680 readings.
To ensure that no data is lost, once the memory is full the thermometer will stop logging data (data will not be overwritten).While recording, the following information is displayed: Current, Minimum and Maximum temperature achieved and cumulative running time. Once an alarm condition has occurred, the following additional information is displayed: Alarm Tally, ALARM notification, cumulative alarm time.
Individual Logger-Trac Thermometers are pre-programmed with alarm values of 2 °C (low) and 8 °C (high) and a recording interval to allow for 30 days of recording (readings are logged every 5 minutes, 33 seconds).
Specifications
TypeDatalogger
Display TypeLCD
Data Memory7680 Readings
CalibrationNot Calibrated
Alarm TypesHigh/Low
Software TypeTraceableLIVE
DescriptionWith temperature MIN/MAX memory. data logging and exportable data
InterfaceUSB
IP RatingIP66
Sensor TypesInternal Sensor
For Use With (Application)Perfect solution for monitoring material in transit as well as reagents, clinical samples, vaccines, pharmaceuticals and food items in any location
Accuracy (Temperature)±0.2°C (0°C; to 10°C;), ±1°C otherwise
Temperature Range (English)-31°F to +122°F
Temperature Range (Metric)-35°C to +50°C
Dimensions (L x W x H)2 x 1 x 0.5 in.
Unit SizeEA