Traceable™ Refrigerator/Freezer Ultra™ Thermometers
Traceable™ Refrigerator/Freezer Ultra™ Thermometers
Actual product may vary
Traceable™ Refrigerator/Freezer Ultra™ Thermometers
Traceable™ Refrigerator/Freezer Ultra™ Thermometers are designed to meet current CDC requirements for vaccine storage and monitoring.
Have Questions?
Catalog NumberTemperature Range (Metric)
CTL14-650-106-50°C to +70°C
Catalog number CTL14-650-106
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Temperature Range (Metric):
-50°C to +70°C
Traceable™ Refrigerator/Freezer Ultra™ Thermometers are designed to meet current CDC requirements for vaccine storage and monitoring.
  • Triple display simultaneously shows min., max. and current temperatures for a specified probe
  • Smart-Alarm™ with visual and audible indicators signal continuously when an out-of-range condition is sensed
  • Alarm continues even after unit returns to non-alarm conditions and is programmable in 0.1° increments
  • °C/°F switchable
  • Enclosed temperature-buffered sensor to accurately monitor temperatures in one or two different areas in refrigerators, freezers, water baths and incubators
  • Solid-state probe sealed in a miniature bottle (1 x 2.5 in.) eliminates mercury contamination and rapid temperature changes when refrigerator door is opened
  • Miniature bottle is filled with nontoxic glycol, generally recognized as safe (GRAS) by FDA thereby eliminating concerns about contact with food or drinking water
  • Features high-impact, chemical-resistant ABS plastic case (2.75 x 4.25 x 0.75 in.)
  • Ten-foot micro-cable permits refrigerator doors to close on it
  • Range: -50°C to +70°C (-58°F to +158°F)
  • Accuracy: ±0.5°C between -30°C to +50°C, otherwise ±1°C
  • Resolution: 0.1°
Specifications
Bottle FillNontoxic Glycol
Bottle Size2.5 x 6.3 cm (1 x 2.5 in.)
Certifications/ComplianceNIST Traceable
Dimensions (L x W x H)7 x 10.8 x 2 cm (2.75 x 4.25 x 0.75 in.)
For Use With (Application)Monitoring Temperatures in Freezers, Water Baths, Heating Blocks, Incubators, and Refrigerators
IncludesTraceable™ Certificate, Flip-open Stand, 10 ft. Micro Cable, Wall Mounting, Velcro™, Magnetic Strips
Length (Metric)7 cm
MaterialABS Plastic
Probe TypeTwo Bullet Probes
Resolution0.1°
Weight (English)4 oz.
Weight (Metric)113.4 g
Product TypeRefrigerator/Freezer Ultra Thermometer
Max. Temperature (English)158°F
Max. Temperature (Metric)70°C
Min. Temperature (English)-58°F
Min. Temperature (Metric)-50°C
Temperature Range (English)-58°F to +158°F
Temperature Range (Metric)-50°C to +70°C
CalibratedYes
For Use With (Equipment)Monitoring Temperature in Freezer, Water Bath, Heating Block, Incubator and Refrigerator
Accuracy±0.5°C between -30°C to +50°C, otherwise ±1°C
DisplayDigital
Unit SizeEA