Traceable™ Four-Channel Countdown Alarm Digital Timer/Stopwatch with Memory Recall
Actual product may vary
Traceable™ Four-Channel Countdown Alarm Digital Timer/Stopwatch with Memory Recall
Traceable™ Four-Channel Countdown Alarm Digital Timer/Stopwatch with Memory Recall
Traceable™ Four-Channel Countdown Alarm Digital Timer/Stopwatch with Memory Recall
Traceable™ Four-Channel Countdown Alarm Digital Timer/Stopwatch with Memory Recall
Traceable™ Four-Channel Countdown Alarm Digital Timer/Stopwatch with Memory Recall
Traceable™ Four-Channel Countdown Alarm Digital Timer/Stopwatch with Memory Recall
Traceable™ Four-Channel Countdown Alarm Digital Timer/Stopwatch with Memory Recall
Traceable™ Four-Channel Countdown Alarm Digital Timer/Stopwatch with Memory Recall
Traceable™ Four-Channel Countdown Alarm Digital Timer/Stopwatch with Memory Recall
Traceable™ Four-Channel Countdown Alarm Digital Timer/Stopwatch with Memory Recall
Traceable™ Four-Channel Countdown Alarm Digital Timer/Stopwatch with Memory Recall
Four countdown channels plus stopwatch channel and time-of-day clock
Have Questions?
Catalog NumberTime Range
CTL14-649-1799 hr. 59 min. 59 sec.
Catalog number CTL14-649-17
Price (SGD)
53.00
Special offer
Ends: 31-Mar-2026
94.00
Save 41.00 (44%)
Each
Time Range:
99 hr. 59 min. 59 sec.
Price (SGD)
53.00
Special offer
Ends: 31-Mar-2026
94.00
Save 41.00 (44%)
Each
Fisher Scientific™ Traceable™ Four-Channel Countdown Alarm Timer/Stopwatch with Memory Recall features four countdown channels plus stopwatch channel and time-of-day clock.
Timer: Counts down from 99 hours, 59 minutes, 59 seconds to 1 second. Stopwatch: Counts up from 1 second to 99 hours, 59 minutes, 59 seconds. 
  • Timing capacity: 100 hours
  • Accuracy: 0.01%
  • “Thru-zero” function counts down, alarms, counts up
  • Includes as 12/24 hour clock
  • Time-out mode
  • Easel stand, lapel clip, hole for lanyard and magnetic back
  • Alarm can be silenced automatically or manually
  • Extra-loud, 70 db alarms for 1 minute or can be silenced manually
  • Memory recall
  • Time-of-day clock feature
  • Dimensions: 6.35L x 6.03W x 1.27cmH (2.50 x 2.375 x 0.5 in.)
Specifications
Product TypeDigital Stopwatch/Timer
Time Range99 hr. 59 min. 59 sec.
AlarmsAudible
Accuracy±0.01%
Resolution1 sec.
DisplayLCD
Dimensions (L x W x H)2.75 x 2.5 x 0.5 in. (7 x 6.4 x 1.3 cm)
Length (English)2.75 in.
Length (Metric)7 cm
Width (English)2.5 in.
Width (Metric)6.4 cm
Height (English)0.5 in.
Height (English) Display0.75 in.
Height (Metric)1.3 cm
Height (Metric) Display1.91 cm
Weight (English)2 oz.
Weight (Metric)56.69 g
IncludesEasel stand, lapel clip, hole for lanyard and magnetic back, two-year silver-oxide battery, sturdy ABS plastic case
For Use With (Application)High display that permits viewing from across the lab
Certifications/ComplianceISO 17025, A2LA, NIST
Unit SizeEach