Traceable™ Countdown/Countup Timer Stopwatch
Actual product may vary
Traceable™ Countdown/Countup Timer Stopwatch
Convenient, one-channel benchtop timer.
Have Questions?
Catalog NumberTime Range
CTL14-649-1499 min. 59 sec. to 1 sec.
Catalog number CTL14-649-14
Price (SGD)
87.00
Each
Time Range:
99 min. 59 sec. to 1 sec.
Price (SGD)
87.00
Each
Convenient, one-channel benchtop timer.
  • Timing Capacity: Countdown from 100 min.; countup to 60 min.
  • Accuracy: 0.01%
  • Resolution: 1 sec.
  • Display: 0.375 in.H LCD
  • Thru-zero function
  • Full keyboard (0 to 9 keys)
  • Alarm: One minute, silenced automatically or manually
  • Spring fastener slip, hole for use with a lanyard, clock and memory
  • Dimensions: 5.71 x 6.35 x 1.27cm (2.25 x 2.5 x 0.5 in.)
Specifications
Product TypeHandheld/Benchtop Timer
Time Range99 min. 59 sec. to 1 sec.
Alarms1 min.
Accuracy0.0001
DisplayLCD
Battery TypeD357/357A
Dimensions (L x W x H)2.25 x 2.5 x 0.5 in. (5.71 x 6.35 x 1.27 cm)
Length (English)2.25 in.
Length (Metric)5.71 cm
Width (English)2.5 in.
Width (Metric)6.35 cm
Height (English)0.5 in.
Height (English) Display0.37 in.
Height (Metric)1.27 cm
Height (Metric) Display0.93 cm
Weight (English)1.5 oz.
Weight (Metric)42.52 g
IncludesMagnetic back, flip open easel stand, spring fastener slip, hole for use with a lanyard, Silver Oxide battery, clock and memory, ABS plastic case
Certifications/ComplianceNIST
Unit SizeEach