Traceable™ Waterproof Food Thermometer with Holders, Food thermometer
Traceable™ Waterproof Food Thermometer with Holders, Food thermometer
Actual product may vary
Traceable™ Waterproof Food Thermometer with Holders, Food thermometer
Unique holder allows piercing probe to be positioned at any height while attached to beakers, cylinders, or vats
Have Questions?
Catalog NumberProduct TypeAccuracy
CTL14-649-100Food Thermometer±1.5°C
Catalog number CTL14-649-100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Product Type:
Food Thermometer
Accuracy:
±1.5°C
Unique holder allows piercing probe to be positioned at any height while attached to beakers, cylinders, or vats. With the touch of a key, Fisher Scientific™ Traceable™ Waterproof Food Thermometer displays the min. and max. temperatures over any period of time.
  • Range is -50 to +300°C (-58 to +572°F)
  • Resolution is 0.1° from -20 to +200° (1° otherwise)
  • Accuracy is ±1.5°C or ±0.4°C at tested points for Ultra™ Model
  • Size: stem length, 3.5 in.; stem diameter,0.14 in.; overall length, 7.25 in.
Specifications
Certifications/ComplianceNIST
Diameter (English) Probe0.14 in.
For Use With (Application)Food labs, food processing, food preparation and brewing
IncludesStainless Steel probe, probe holder, key chain and lanyard
Length (English) Probe3.5 in.
Length (Metric)184 mm
Length (Metric) Probe8.8 cm
MaterialABS Plastic
Resolution0.1°C
Weight (English)1 oz.
Weight (Metric)28.4 g
Product TypeFood Thermometer
Length (English)7.25 in.
Max. Temperature (English)572°F
Max. Temperature (Metric)300°C
Min. Temperature (English)-58°F
Min. Temperature (Metric)-50°C
Temperature Range (English)-58°F to +572°F
Temperature Range (Metric)-50°C to +300°C
CalibratedYes
Accuracy±1.5°C
DisplayDigital LCD
Width (Metric)8 cm
Unit SizeEA