Traceable™ Clock/Thermometer/Calendar
Traceable™ Clock/Thermometer/Calendar
Actual product may vary
Traceable™ Clock/Thermometer/Calendar
Unique clock combination shows the time, month, day, date, and temperature
Have Questions?
Catalog NumberDisplay
CTL14-648-5LCD
Catalog number CTL14-648-5
Price (SGD)
237.00
EA
Add to cart
Display:
LCD
Price (SGD)
237.00
EA
Add to cart
With a huge 2.13 in.H (5.4cm) LCD visible from 35 ft. (10.7m), the Fisher Scientific™ Traceable™ Clock/Thermometer/Calendar features a stylish two-tone color face and slim silhouette.
  • Wall-mounted, battery-operated perpetual clock
  • Timing accuracy: 0.01%
  • Internal temperature sensor displays current reading in your choice of °C (-5° to +55°C range) or °F (23° to 131°F)
Specifications
Product TypeDigital Clock
Accuracy0.0001 sec.
Resolution1 sec.
DisplayLCD
Battery TypeAA
Temperature Range (English)23°F to 122°F
Temperature Range (Metric)-5°C to +50°C
Temperature Resolution0.5°
Dimensions (L x W)9.25 x 10 in. (23.5 x 25.4 cm)
Length (English)9.25 in.
Length (Metric)23.5 cm
Width (English)10 in.
Width (Metric)25.4 cm
Depth (English)1 in.
Depth (Metric)2.54 cm
Weight (English)19 oz.
Weight (Metric)0.53 kg
IncludesBatteries (2 AA), wall mount
For Use With (Application)High tech design looks great in any science classroom or office
QuantityEach
Certifications/ComplianceISO 17025, A2LA, NIST
Unit SizeEA