Traceable™ Temperature Chart Recorders
Actual product may vary
Traceable™ Temperature Chart Recorders
Provides a permanent record, which is easy to read, duplicate, fax, scan and file. Traceable™ Temperature Chart Recorders are single-use, disposable recorders for monitoring temperatures in refrigerators, supply rooms, food storage, chemical warehouses, animal quarters, incubators, goods-in-transit, walk-in chambers, biological materials, and manufacturing plants.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
CTL14-648-4110-Day Recorder
CTL14-648-4240-Day Recorder
Catalog number CTL14-648-41
Price (SGD)
144.00
Each
Product Type:
10-Day Recorder
Price (SGD)
144.00
Each
Provides a permanent record, which is easy to read, duplicate, fax, scan and file. Traceable™ Temperature Chart Recorders are single-use, disposable recorders for monitoring temperatures in refrigerators, supply rooms, food storage, chemical warehouses, animal quarters, incubators, goods-in-transit, walk-in chambers, biological materials, and manufacturing plants.
  • Shock-resistant ABS plastic case
  • Eliminate messy pens, expensive instrumentation, confusing software and training
  • Display temperature in both °F and °C on large 5.5 in. dia. (14 cm) chart
  • Battery-operated motor turns the chart with accuracy of ±1 second per day
  • Easy-to-read chart shows temperature and time/day of occurrence
  • Transparent window on recorder allows viewing of current temperature and history
  • Records for 5, 10 or 40 days
  • Chart, marker, and battery are preloaded—simply pull tab to begin recording
  • Tamper-evident seal maintains integrity of unit and results
  • Case measures 3.87 L x 2.5 W x 5.25 in. H (13.3 x 9.8 x 6.4 cm)
Specifications
Accuracy±1°C (±2°F)
Certifications/ComplianceISO 17025, NIST
Battery TypeAA Alkaline
Product Type10-Day Recorder
Temperature Range (English)-20°F to +100°F
Height (English)2.5 in.
Height (Metric)6.4 cm
IncludesOne AA Alkaline Battery
Rotational Speed5 Days
SensorBimetallic
Temperature Range (Metric)-28°C to +38°C
Weight (English)11 oz.
Weight (Metric)312 g
Width (English)3.87 in.
Width (Metric)9.8 cm
Unit SizeEach