Traceable™ Temperature/Humidity Meter
Traceable™ Temperature/Humidity Meter
Actual product may vary
Traceable™ Temperature/Humidity Meter
Simultaneously displays humidity, dew point and temperature
Have Questions?
Catalog NumberTemperature Range (Metric)
CTL11-661-210°C to 50°C
Catalog number CTL11-661-21
Price (SGD)
-
Temperature Range (Metric):
0°C to 50°C
Simultaneously displays humidity, dew point and temperature. Fisher Scientific™ Traceable™ Temperature/Humidity Meter are ideal for use in cleanrooms, food-processing facilities, laboratories, and other humidity-sensitive areas.
  • Serial RS-232 output allows connection to a computer to download datalogger results for constant monitoring
  • Datalogger captures 1000 readings manually or automatically when set between 1 and 3600 second (sold separately)
  • Temperature range, 0° to 50°C (32° to 122°F), with 0.01° resolution, ±1°C accuracy
  • Relative humidity range, 10 to 95% RH, with 0.01% resolution, ±3% accuracy
  • Memory recalls maximum, minimum, and average readings
  • HOLD button freezes display for later reading
  • Large display (1.37 in.H) shows temperature and humidity simultaneously
Specifications
TypeThermohygrometer
Humidity Range10 to 95%
Resolution0.0001
Accuracy±3%
Temperature Range (Metric)0°C to 50°C
Temperature Accuracy±1°C
Display TypeLCD
MaterialABS Plastic
MemoryMinimum/maximum readings
Temperature Range (English)32°F to 122°F
Temperature Resolution0.01°F
Dimensions (L x W x H)3 x 1.25 x 7 in. (7.6 x 3.2 x 17.8 cm)
Length (English)3 in.
Length (English) Cable40 in.
Length (English) Probe8 in.
Length (Metric)7.6 cm
Length (Metric) Cable101.6 cm
Length (Metric) Probe20.32 cm
Width (English)1.25 in.
Width (Metric)3.2 cm
Height (English)7 in.
Height (Metric)17.8 cm
Diameter (English) Probe0.66 in.
Diameter (Metric) Probe1.67 cm
Weight (English)9.5 oz.
Weight (Metric)0.269 kg
For Use With (Application)Ideal for use in cleanrooms, food-processing facilities, laboratories and other humidity-sensitive areas
Includes0.66 dia. x 8 in. L probe, 40 in. cable and 9V alkaline battery
Certifications/ComplianceISO 17025, A2LA, NIST
Unit SizeEach