▸
pH and Electrochemistry
▸
Probes and Electrodes
▸
Conductivity Electrodes
▸
Replacement Probes for Traceable™ Conductivity and TDS Meters
Actual product may vary
Replacement Probes for Traceable™ Conductivity and TDS Meters
For use with Traceable Conductivity and TDS Meters.
Have Questions?
Catalog Number
Model
CTL09-326-2A
For 09-326-2 Meter
Catalog number
CTL09-326-2A
Price (SGD)
753.00
Each
Add to cart
Model:
For 09-326-2 Meter
Price (SGD)
753.00
Each
Add to cart
For use with Traceable Conductivity and TDS Meters.
Specifications
For Use With (Equipment)
Traceable Conductivity/TDS meter 09-326-2
Type
Conductivity Probe
Connector Types
BNC
Material
Glass
Measurement Ranges
0.05 to 200,000 μomhos
Model
For 09-326-2 Meter