Traceable™ Water-Resistant Timer
Actual product may vary
Traceable™ Water-Resistant Timer
Ideal for wet labs, washdown areas, plants, and field use
Have Questions?
Catalog NumberDisplay
CTL06-664-40LED
Catalog number CTL06-664-40
Price (SGD)
-
Display:
LED
Fisher Scientific™ Traceable™ Water-Resistant Timer is ideal for wet labs, washdown areas, plants, and field use.
  • Times from 99 minutes, 59 seconds to zero with one-second resolution
  • Accuracy: 0.01%
  • Splashproof
  • When zero is reached, a bright-red LED flashes as a visual sign
  • Audible alarm sounds for one minute or until manually turned off
  • Unit also features infinite time-outs
Specifications
Product TypeWater-resistant Timer
Accuracy0.0001
Resolution1 sec.
DisplayLED
Battery TypeSilver Oxide
Diameter (English) Inner3 in.
Diameter (Metric) Inner7.62 cm
Weight (English)1.5 oz.
Weight (Metric)42.52 g
IncludesLanyard, Magnet, Velcro™ (to affix to surfaces), Battery
Certifications/ComplianceNIST
Unit SizeEach