▸
Buffers and Standards
▸
Electrochemistry Buffers
▸
Traceable™ pH Buffer Standards, Certified Reference Material (CRM), Fisherbrand™
Element Search
Structure Search
Traceable™ pH Buffer Standards, Certified Reference Material (CRM), Fisherbrand™
IUPAC formulation
Have Questions?
Change view
Quantity:
6 x 100 mL
16 oz.
Packaging:
View all
6 shots of 100mL
pH:
View all
4.005
7.00
10.012
Catalog number
CTL06-664-262
Price (SGD)
74.00
Pack
Add to cart
Quantity:
6 x 100 mL
Packaging:
6 shots of 100mL
pH:
4.005
Request bulk or custom format
Price (SGD)
74.00
Pack
Add to cart
Specifications
Chemical Name or Material
pH Buffer Standards
ChemAlert Storage Symbol
Gray
For Use With (Application)
Calibration
For Use With (Equipment)
Instruments and Electrodes
pH
4.005
View more
IUPAC formulation
Compatible with all makes and instruments and electrodes
Accuracy at 25°C is ±0.010 pH