Traceable™ Vaccine Refrigerator/Freezer Thermometer
Actual product may vary
Traceable™ Vaccine Refrigerator/Freezer Thermometer
Traceable™ Vaccine Refrigerator/Freezer Thermometer
Traceable™ Vaccine Refrigerator/Freezer Thermometer
Traceable™ Vaccine Refrigerator/Freezer Thermometer
Traceable™ Vaccine Refrigerator/Freezer Thermometer
Traceable™ Vaccine Refrigerator/Freezer Thermometer
Traceable™ Vaccine Refrigerator/Freezer Thermometer
Traceable™ Vaccine Refrigerator/Freezer Thermometer
Traceable™ Vaccine Refrigerator/Freezer Thermometer
Traceable™ Vaccine Refrigerator/Freezer Thermometer
Traceable™ Vaccine Refrigerator/Freezer Thermometer
Monitors temperature of vaccine in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberBottle Size
CTL06-664-1110 mL
CTL15-077-7205 mL
Catalog number CTL06-664-11
Price (SGD)
115.70
Special offer
Ends: 06-Sep-2026
178.00
Save 62.30 (35%)
Each
Bottle Size:
10 mL
Price (SGD)
115.70
Special offer
Ends: 06-Sep-2026
178.00
Save 62.30 (35%)
Each
Monitors temperature of vaccine in freezers, water baths, heating blocks, incubators, and refrigerators with enclosed temperature-buffered sensor. Fisherbrand™ Traceable™ Vaccine Refrigerator/Freezer Thermometer includes a probe which is sealed in glycol solution and insulates sensor when refrigerator door is opened.
  • Triple display simultaneously shows Min/max and current temperatures, °F/°C switchable
  • Bottle insulates sensor from rapid temperature changes when refrigerator door is opened
  • 15-077-720 has a 5mL bottle designed for vaccines; 06-664-11 has a 10mL bottle
  • Glass bottle filled with glycol mimics vaccine temperature and fits in trays
  • Probe is sealed in glycol solution which insulates sensor when refrigerator door is opened
  • Ten-foot micro-cable permits refrigerator doors to close on it
  • Minimum/maximum memory monitors high and low readings overnight, on weekends, or for any time period
  • Alarm is programmable in 1° increments, sounds continuously when temperature rises above/falls below set points and can be turned off manually
  • High-impact, chemical-resistant ABS plastic case
  • Temperature range: -50 to +70°C/-58 to +158°F
  • Resolution: 1° accuracy: ±1C
  • Battery:AA Alkaline (1)
Specifications
Bottle FillGlycol
Bottle Size10 mL
Certifications/ComplianceISO 17025, A2LA, NIST
Dimensions (L x W x H)2.75 x 2.75 x 0.75 in. (69.85 x 70 x 19 mm)
For Use With (Application)Monitoring temperature in freezers, water baths, heating blocks, incubators and refrigerators
Height (English)0.75 in.
Height (Metric)19 mm
IncludesVelcro™ and magnetic mounting strip, one AA battery, ABS Plastic case, alarm set in 1° increments alerts user with visual and audible signals if temperature goes out
Length (English) Cable10 in.
Length (Metric)70 mm
Length (Metric) Cable25.4 cm
MaterialABS Plastic
ModelRefrigerator/Freezer Thermometer with 10mL bottle
Resolution1°C
Weight (English)4 oz.
Weight (Metric)0.11 kg
Width (English)2.75 in.
Product TypeFridge/Freezer Digital Thermometer
Length (English)2.75 in.
Max. Temperature (English)158°F
Max. Temperature (Metric)70°C
Min. Temperature (English)-58°F
Min. Temperature (Metric)-50°C
Temperature Range (English)-58°F to +158°F
Temperature Range (Metric)-50°C to +70°C
CalibratedYes
For Use With (Equipment)Refrigerators/Freezers
Battery TypeAA Alkaline (1)
Accuracy±1°C
DisplayDigital LCD
Width (Metric)70 mm
Unit SizeEach