Specifications
TypeClock Thermometer/Humidity Thermometer
Humidity Range0 to 100%
Resolution0.02
Accuracy±5%
Temperature Range (Metric)-20°C to +55°C
Temperature Accuracy±1.5°C
Temperature Range (English)10°F to 130°F
Temperature Resolution2°F
Weight (English)1 lb.
Weight (Metric)0.453 kg
For Use With (Application)Keep track of the current time, temperature and humidity all at once
IncludesAA battery and wall mount screw
Certifications/ComplianceISO 17025, NIST
Unit SizeEach