Traceable™ Digital Three-Channel Alarm Timer with Triple-Line LCD
Actual product may vary
Traceable™ Digital Three-Channel Alarm Timer with Triple-Line LCD
Traceable™ Digital Three-Channel Alarm Timer with Triple-Line LCD
Traceable™ Digital Three-Channel Alarm Timer with Triple-Line LCD
Traceable™ Digital Three-Channel Alarm Timer with Triple-Line LCD
Traceable™ Digital Three-Channel Alarm Timer with Triple-Line LCD
Traceable™ Digital Three-Channel Alarm Timer with Triple-Line LCD
Traceable™ Digital Three-Channel Alarm Timer with Triple-Line LCD
Traceable™ Digital Three-Channel Alarm Timer with Triple-Line LCD
Traceable™ Digital Three-Channel Alarm Timer with Triple-Line LCD
Traceable™ Digital Three-Channel Alarm Timer with Triple-Line LCD
Accuracy of 0.001%
Have Questions?
Catalog NumberTime Range
CTL06-662-599 hr. 59 min. 59 sec.
Catalog number CTL06-662-5
Price (SGD)
176.00
Each
Time Range:
99 hr. 59 min. 59 sec.
Price (SGD)
176.00
Each
Fisher Scientific™ Traceable™ Three-Channel Alarm Timer with Triple-Line LCD offers three-channel timing with individual or simultaneous start.
  • Three-line display
  • One-second resolution
  • “Thru-zero” function counts down, alarms and counts up
  • Counts down and up simultaneously
  • Time-out mode
  • Memory recall automatically returns to preset time
  • Alarm volume is adjustable
  • Visual and audible alarm operates for one minute or continuously
  • Time-of-day clock with AM/PM indicators
  • Four rubber feet keep instrument off wet bench
  • LCD is 0.75 in.H (9.5mm)
  • Timing range: 99 hours, 59 minutes, 59 seconds to 1 second
  • Stopwatch: 1 second to 99 hours, 59 minutes, 59 seconds
  • Accuracy: 0.001%
Specifications
Product TypeHandheld/Benchtop Timer
Time Range99 hr. 59 min. 59 sec.
AlarmsAudible/Visible
Accuracy1e-005
Resolution1 sec.
DisplayLCD
Dimensions (L x W x H)1 x 3 x 3.25 in. (2.54 x 7.62 x 8.22 cm)
Length (English)1 in.
Length (Metric)2.54 cm
Width (English)3 in.
Width (Metric)7.62 cm
Height (English)3.25 in.
Height (Metric)8.22 cm
Weight (English)3 oz.
Weight (Metric)85.04 g
IncludesAAA alkaline battery for up to +two years of use and chemical-resistant ABS plastic case.
Certifications/ComplianceISO 17025, A2LA, NIST
Unit SizeEach