Traceable™ Lab-Top Timer
Traceable™ Lab-Top Timer
Actual product may vary
Traceable™ Lab-Top Timer
Lab-Top Timer is built expressly for bench use
Have Questions?
Catalog NumberDisplay
CTL06-662-45LCD
Catalog number CTL06-662-45
Price (SGD)
-
Display:
LCD
Lab-Top Timer is built expressly for bench use. Ease of operation, versatile timing modes, extra-large keys, and the any-angle viewing make Fisherbrand™ Traceable™ Lab-Top Timer an ideal unit for the lab.
  • LCD screen is designed for viewing at any angle (sitting or standing)
  • Counts down from 24 hours to 1 second, in second increments, with an accuracy of 0.01%
  • Special memory chip allows programming an instant replay repeat time which appears on the display at the touch of a key
  • Memory time is locked in until changed
  • Count-up (stopwatch) mode and a time-of-day clock
  • Size: 3.25 x 3.25 x 1.75 in.
  • Weight: 4 oz.
Specifications
Product TypeHandheld/Benchtop Timer
Time Range24 hr. to 1 sec.
AlarmsAudible/Visible
Accuracy0.0001
Resolution1 sec.
DisplayLCD
Battery TypeAA
Weight (English)4 oz.
Weight (Metric)113.39 g
IncludesIndividually serial-numbered Traceable Certificate and a 3-year battery
Certifications/ComplianceNIST
Unit SizeEA