Tight-Seal™ Clamp Assortment
Tight-Seal™ Clamp Assortment
Actual product may vary
Tight-Seal™ Clamp Assortment
Tight-Seal™ Clamp Assortment
Tight-Seal™ Clamp Assortment
Kit ensures that proper size is always handy
Have Questions?
Catalog number CTL05-815
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Kit ensures that proper size is always handy
  • With five each of the 13 sizes of Fisherbrand Tight-Seal clamps
Specifications
TypeHose and Tubing Clamp
Includes65 clamps, 5 of each 13 sizes
Diameter (English) Inside0.24 to 1.61 in.
Temperature (Metric) Resistant0°C to 90°C
Temperature (English) Resistant32°F to 194°F
Diameter (Metric) Inside6.25 to 40.90 mm
Unit SizeEA